6-Amino-1-hexanol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Aminohexan-1-ol

L14157.06
L14157.06EA 60.9 EUR
L14157.06
6-Amino-1-hexanol 97%
6-Amino-1-hexanol

Purity: 97% CAS No.: 4048-33-3 Molecular Formula: H2N(CH2)6OH Formula Weight: 117.19 Flash Point: 133°(271°F) Melting Point: 55-58° Boiling Point: 136-139°/30mm Beilstein Reference: 1732524 MDL No.: MFCD00008241

Formel: C₆H₁₅NO
Molecular Weight: 117,19 g/mol
Siedepunkt: 136…139 °C (30 mmHg)
Schmelzpunkt: 55…58 °C
Flammpunkt: 133 °C (271 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008241
CAS-Nummer: 4048-33-3
EINECS: 223-748-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR