3',4'-Dihydroxyflavon ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(3,4-Dihydroxyphenyl)chromen-4-on, 2-(3,4-Dihydroxyphenyl)-4-benzopyron

L14161.MC
L14161.MCEA 64.7 EUR
L14161.MC
3',4'-Dihydroxyflavon ≥97%
3',4'-Dihydroxyflavon

Purity: 97% CAS No.: 4143-64-0 Molecular Formula: C15H10O4 Formula Weight: 254.24 Melting Point: 249-252° Beilstein Reference: 212540

Formel: C₁₅H₁₀O₄
Molecular Weight: 254.24 g/mol
Schmelzpunkt: 249…252 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017600
CAS-Nummer: 4143-64-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR