4-(Dimethylamino)benzoylchlorid 97%

Lieferant: Thermo Fisher Scientific
Gefahr

L14176.06 L14176.14
L14176.06EA 42.2 EUR
L14176.06 L14176.14
4-(Dimethylamino)benzoylchlorid 97%
4-(Dimethylamino)benzoylchlorid

Purity: 97% CAS No.: 4755-50-4 Molecular Formula: C9H10ClNO Formula Weight: 183.64 Melting Point: 142-148° Beilstein Reference: 1100858 UN No.: 3261 MDL No.: MFCD00236039

Formel: C₉H₁₀ClNO
Siedepunkt: 180 °C (15 mmHg)
Schmelzpunkt: 142…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236039
CAS-Nummer: 4755-50-4
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR