2,4,6-Trichlorbenzoesäure ≥94%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,4,6-TBA

L14191.06
L14191.06EA 43.6 EUR
L14191.06
2,4,6-Trichlorbenzoesäure ≥94%
2,4,6-Trichlorbenzoesäure

Purity: 94% CAS No.: 50-43-1 Molecular Formula: C7H3Cl3O2 Formula Weight: 225.46 Melting Point: 162-164° Beilstein Reference: 1874127

Formel: C₇H₃Cl₃O₂
Molecular Weight: 225,46 g/mol
Schmelzpunkt: 162…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060699
CAS-Nummer: 50-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR