(S)-(+)-Epichlorhydrin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98+%
CAS No.: 67843-74-7
Molecular Formula: C3H5ClO
Formula Weight: 92.53
Storage Temperature: 2°C to 8°C
Flash Point: 32°(90°F)
Refractive Index: 1.4380
Boiling Point: 116-117°
Beilstein Reference: 1420784
Optical Rotation: +35° (c=1 in methanol)
UN No.: 2023
MDL No.: MFCD00077760
Formel:
ClCH₂CH(O)CH₂ Siedepunkt: 116…117 °C Schmelzpunkt: –48 °C Dichte: 1,180 Flammpunkt: 32 °C (90 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00077760 CAS-Nummer: 67843-74-7 UN: 2023 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...