(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(-)-MTPA-Cl, (-)-Mosher's Acid Chloride, (R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
L14325.MF
L14325.MD
L14325.03
L14325.MFEA
46.2
EUR
L14325.MF
L14325.MD
L14325.03
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Purity: 98+%
CAS No.: 39637-99-5
Molecular Formula: C10H8ClF3O2
Formula Weight: 252.62
Storage Temperature: -30°C to -10°C
Flash Point: 89°(192°F)
Refractive Index: 1.4690
Boiling Point: 213-214°
Beilstein Reference: 3591565
Optical Rotation: -137° (neat)
UN No.: UN3265
MDL No.: MFCD00044400
Formel:
C₁₀H₈ClF₃O₂ Molecular Weight: 252.62 g/mol Siedepunkt: 213…214 °C Dichte: 1.353 Flammpunkt: 89 °C (192 °F) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00044400 CAS-Nummer: 39637-99-5 UN: 3265 ADR: 8 II Merck Index: 14,6280 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...