Bis(triphenylphosphoranylidene)ammonium chloride, (max. 3% H₂O) 97% (Trockengewicht)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L14339.06
L14339.14
L14339.06EA
76.5
EUR
L14339.06
L14339.14
Bis(triphenylphosphoranylidene)ammonium chloride, (max. 3% H₂O) 97% (Trockengewicht)
Bis(triphenylphosphoranylidene)ammonium chloride
Purity: 97%
CAS No.: 21050-13-5
Molecular Formula: C36H30ClNP2
Formula Weight: 574.05
Melting Point: 274-276°
Beilstein Reference: 4097979
MDL No.: MFCD00151523
Formel:
C₃₆H₃₀ClNP₂ Schmelzpunkt: 274…276 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151523 CAS-Nummer: 21050-13-5 EINECS: 244-170-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...