Bis(triphenylphosphoranylidene)ammonium chloride, (max. 3% H₂O) 97% (Trockengewicht)

Lieferant: Thermo Fisher Scientific
Achtung

L14339.06 L14339.14
L14339.06EA 76.5 EUR
L14339.06 L14339.14
Bis(triphenylphosphoranylidene)ammonium chloride, (max. 3% H₂O) 97% (Trockengewicht)
Bis(triphenylphosphoranylidene)ammonium chloride

Purity: 97% CAS No.: 21050-13-5 Molecular Formula: C36H30ClNP2 Formula Weight: 574.05 Melting Point: 274-276° Beilstein Reference: 4097979 MDL No.: MFCD00151523

Formel: C₃₆H₃₀ClNP₂
Schmelzpunkt: 274…276 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151523
CAS-Nummer: 21050-13-5
EINECS: 244-170-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR