Triethylammoniumdihydrogentrifluorid (∼37% HF)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Triethylamin Trishydrofluorid, Triethylamin Trihydrofluorid

L14417.06 L14417.14 L14417.22
L14417.06EA 28.9 EUR
L14417.06 L14417.14 L14417.22
Triethylammoniumdihydrogentrifluorid (∼37% HF)
Triethylammonium dihydrogentrifluoride

CAS No.: 73602-61-6 Molecular Formula: C6H15N.3HF Formula Weight: 161.21 Flash Point: 87°(188°F) Refractive Index: 1.3915 Melting Point: -29° Boiling Point: 70°/15mm Beilstein Reference: 5522945 UN No.: 1790 MDL No.: MFCD00043294

Formel: C₆H₁₈F₃N
Molecular Weight: 161.21 g/mol
Siedepunkt: 70 °C (15 mmHg)
Schmelzpunkt: –29 °C
Dichte: 0.990
Flammpunkt: 87 °C (188 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00043294
CAS-Nummer: 73602-61-6
EINECS: 277-550-5
UN: 1790
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR