(R)-(+)-4-Isopropyl-2-oxazolidinon ≥98%
Lieferant: Thermo Scientific
Synonyme:
(R)-4-Isopropyloxazolidin-2-one, (4R)-(+)-4-Isopropyl-2-oxazolidinone
L14518.03
L14518.03EA
103
EUR
L14518.03
(R)-(+)-4-Isopropyl-2-oxazolidinon ≥98%
(R)-(+)-4-Isopropyl-2-oxazolidinon
Purity: 98+%
CAS No.: 95530-58-8
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Melting Point: 70-74°
Beilstein Reference: 5376658
Formel:
C₆H₁₁NO₂ Molecular Weight: 129,16 g/mol Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075172 CAS-Nummer: 95530-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...