N,N-Dimethylcyclohexylamin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L14521.AC
L14521.AE
L14521.ACEA
25.9
EUR
L14521.AC
L14521.AE
N,N-Dimethylcyclohexylamin ≥98%
N,N-Dimethylcyclohexylamin
Purity: 98+%
CAS No.: 98-94-2
Molecular Formula: C8H17N
Formula Weight: 127.23
Flash Point: 39°(102°F)
Refractive Index: 1.4540
Melting Point: -60°
Boiling Point: 160-161°
Beilstein Reference: 1919922
UN No.: 2264
MDL No.: MFCD00003844
Formel:
C₈H₁₇N Siedepunkt: 160…161 °C Schmelzpunkt: –60 °C Dichte: 0,849 Flammpunkt: 39 °C (102 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003844 CAS-Nummer: 98-94-2 EINECS: 202-715-5 UN: 2264 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...