4-(2-Hydroxyhexafluoroisopropyl)benzoic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L14554.03
L14554.06
L14554.03EA
71.8
EUR
L14554.03
L14554.06
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid 97%
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid
Purity: 97%
CAS No.: 16261-80-6
Molecular Formula: C10H6F6O3
Formula Weight: 288.15
Odour: Odourless
Melting Point: 122-126°
Beilstein Reference: 2148278
MDL No.: MFCD00236043
Formel:
C₁₀H₆F₆O₃ Schmelzpunkt: 122…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236043 CAS-Nummer: 16261-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...