Methyl-4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate 99%

Lieferant: Thermo Scientific
Achtung

L14622.06
L14622.06EA 63.2 EUR
L14622.06
Methyl-4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate 99%
Methyl-4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate

Purity: 99% CAS No.: 39493-62-4 Molecular Formula: C9H12O4 Formula Weight: 184.19 Melting Point: 122-124° Beilstein Reference: 5258501 MDL No.: MFCD01075661

Formel: C₉H₁₂O₄
Molecular Weight: 184,19 g/mol
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075661
CAS-Nummer: 39493-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR