1-Benzyl-4-Boc-piperazine 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L14644.06
L14644.06EA
166
EUR
L14644.06
1-Benzyl-4-Boc-piperazine 99%
1-Benzyl-4-Boc-piperazine
Purity: 99%
CAS No.: 57260-70-5
Molecular Formula: C16H24N2O2
Formula Weight: 276.38
Melting Point: 71-73°
Beilstein Reference: 797033
MDL No.: MFCD00075603
Formel:
C₁₆H₂₄N₂O₂ Schmelzpunkt: 71…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075603 CAS-Nummer: 57260-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...