Diethoxy(dimethyl)silan ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Dimethyldiethoxysilan

L14725.18 L14725.30 L14725.0B
L14725.18EA 27.8 EUR
L14725.18 L14725.30 L14725.0B
Diethoxy(dimethyl)silan ≥97%
Diethoxy(dimethyl)silan

Purity: 97% CAS No.: 78-62-6 Molecular Formula: C6H16O2Si Formula Weight: 148.28 Flash Point: 11°(52°F) Refractive Index: 1.3820 Odour: Mild Melting Point: -70° Boiling Point: 111-113° Beilstein Reference: 1736110 UN No.: 2380 MDL No.: MFCD00009068

Formel: C₆H₁₆O₂Si
Molecular Weight: 148.28 g/mol
Siedepunkt: 111…113 °C
Schmelzpunkt: –70 °C
Dichte: 0.865
Flammpunkt: 11 °C (52 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009068
CAS-Nummer: 78-62-6
EINECS: 201-127-6
UN: 2380
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR