4'-Chloro-2,2':6',2''-terpyridine 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L14728.03
L14728.MD
L14728.03EA
154
EUR
L14728.03
L14728.MD
4'-Chloro-2,2':6',2''-terpyridine 98%
4'-Chloro-2,2':6',2''-terpyridine
Purity: 98%
CAS No.: 128143-89-5
Molecular Formula: C15H10ClN3
Formula Weight: 267.72
Melting Point: 148-152°
Beilstein Reference: 3613386
MDL No.: MFCD00191930
Formel:
C₁₅H₁₀ClN₃ Schmelzpunkt: 148…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191930 CAS-Nummer: 128143-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...