Bortribromid 1 M in Dichlormethan
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L14880.AC
L14880.AE
L14880.A1
L14880.ACEA
52
EUR
L14880.AC
L14880.AE
L14880.A1
Bortribromid 1 M in Dichlormethan
Bortribromid
CAS No.: 10294-33-4
Molecular Formula: BBr3
Formula Weight: 250.52
Refractive Index: 1.4340
Merck Reference: 14,1347
UN No.: 3390
MDL No.: MFCD00011312
Formel:
BBr₃ Molecular Weight: 250.52 g/mol Siedepunkt: 91,2 Schmelzpunkt: -46 Dichte: 1.467 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011312 CAS-Nummer: 10294-33-4 EINECS: 233-657-9 UN: 3390 ADR: 6.1 I Merck Index: 14,1347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...