Bortribromid 1 M in Dichlormethan

Lieferant: Thermo Fisher Scientific
Gefahr

L14880.AC L14880.AE L14880.A1
L14880.ACEA 52 EUR
L14880.AC L14880.AE L14880.A1
Bortribromid 1 M in Dichlormethan
Bortribromid

CAS No.: 10294-33-4 Molecular Formula: BBr3 Formula Weight: 250.52 Refractive Index: 1.4340 Merck Reference: 14,1347 UN No.: 3390 MDL No.: MFCD00011312

Formel: BBr₃
Molecular Weight: 250.52 g/mol
Siedepunkt: 91,2
Schmelzpunkt: -46
Dichte: 1.467
Lagertemperatur: Raumtemperatur
MDL: MFCD00011312
CAS-Nummer: 10294-33-4
EINECS: 233-657-9
UN: 3390
ADR: 6.1 I
Merck Index: 14,1347

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR