Sodium bromoacetate 98%

Lieferant: Thermo Fisher Scientific
Gefahr

L14983.06 L14983.14
L14983.06EA 32.7 EUR
L14983.06 L14983.14
Sodium bromoacetate 98%
Sodium bromoacetate

Purity: 98% CAS No.: 1068-52-6 Molecular Formula: BrCH2CO2Na Formula Weight: 160.93 Odour: Odourless Melting Point: 300° UN No.: UN2811 MDL No.: MFCD01311832

Formel: BrCH₂CO₂Na
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01311832
CAS-Nummer: 1068-52-6
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR