Lithiumbis(trimethylsilyl)amid 20% ∼1.06 M in THF / Ethylbenzol

Lieferant: Thermo Scientific
Gefahr

Synonyme: Lithiumbis(trimethylsilyl)amid, HMDS Lithiumsalz, N-Lithiohexamethyldisilazan, HMDS-Li

L15012.AE
L15012.AEEA 85.5 EUR
L15012.AE
Lithiumbis(trimethylsilyl)amid 20% ∼1.06 M in THF / Ethylbenzol
Lithiumbis(trimethylsilyl)amid

Packaged in resealable septum cap bottle


CAS No.: 4039-32-1 Molecular Formula: C6H18LiNSi2 Formula Weight: 167.33 Flash Point: -17°(1°F) Beilstein Reference: 8136861 UN No.: 2924 MDL No.: MFCD00008261

Formel: C₆H₁₈LiNSi₂
Molecular Weight: 167,33 g/mol
Dichte: 0,891
Flammpunkt: –17 °C (1 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008261
CAS-Nummer: 4039-32-1
EINECS: 223-725-6
UN: 2924
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR