3-Thiophenethanol ≥98%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
3-(2-Hydroxyethyl)thiophen, Thiophen-3-ethanol, 2-(3-Thienyl)ethanol
L15138.06
L15138.06EA
84.4
EUR
L15138.06
3-Thiophenethanol ≥98%
3-Thiophenethanol
Purity: 98%
CAS No.: 13781-67-4
Molecular Formula: C6H8OS
Formula Weight: 128.19
Flash Point: 92°(197°F)
Refractive Index: 1.5530
Boiling Point: 110-111°/14mm
Beilstein Reference: 107080
MDL No.: MFCD00009766
Formel:
C₆H₈OS Molecular Weight: 128,19 g/mol Siedepunkt: 110…111 °C (14 mmHg) Dichte: 1,159 Flammpunkt: 92 °C (197 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009766 CAS-Nummer: 13781-67-4 EINECS: 237-434-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...