(Bis(trifluoracetoxy)iod)benzol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L15141.06
L15141.14
L15141.06EA
35.5
EUR
L15141.06
L15141.14
(Bis(trifluoracetoxy)iod)benzol ≥97%
(Bis(trifluoracetoxy)iod)benzol
Purity: 97%
CAS No.: 2712-78-9
Molecular Formula: C10H5F6IO4
Formula Weight: 430.04
Storage Temperature: 2°C to 8°C
Melting Point: 122-125°
Beilstein Reference: 764767
MDL No.: MFCD00009672
Formel:
C₁₀H₅F₆IO₄ Schmelzpunkt: 122…125 °C |
MDL:
MFCD00009672 CAS-Nummer: 2712-78-9 EINECS: 220-308-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...