Rhodium(II)acetat dimer ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)

L15152.04 L15152.MC L15152.ME
L15152.04EA 2172 EUR
L15152.04 L15152.MC L15152.ME
Rhodium(II)acetat dimer ≥98%
Rhodium(II)acetat dimer

Purity: 98+% CAS No.: 15956-28-2 Molecular Formula: C8H12O8Rh2 Formula Weight: 441.99 Odour: Odourless Melting Point: 205° MDL No.: MFCD00003538

Formel: C₈H₁₂O₈Rh₂
Molecular Weight: 441.987 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003538
CAS-Nummer: 15956-28-2
EINECS: 240-084-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR