Rhodium(II)acetat dimer ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)
L15152.04
L15152.MC
L15152.ME
L15152.04EA
2172
EUR
L15152.04
L15152.MC
L15152.ME
Rhodium(II)acetat dimer ≥98%
Rhodium(II)acetat dimer
Purity: 98+%
CAS No.: 15956-28-2
Molecular Formula: C8H12O8Rh2
Formula Weight: 441.99
Odour: Odourless
Melting Point: 205°
MDL No.: MFCD00003538
Formel:
C₈H₁₂O₈Rh₂ Molecular Weight: 441.987 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003538 CAS-Nummer: 15956-28-2 EINECS: 240-084-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...