Montmorillonite

Lieferant: Thermo Fisher Scientific

L15160.30
L15160.30EA 34.3 EUR
L15160.30
Montmorillonite
Montmorillonite

CAS No.: 1318-93-0 Odour: Odourless MDL No.: MFCD00132796

Lagertemperatur: Raumtemperatur
MDL: MFCD00132796
CAS-Nummer: 1318-93-0
EINECS: 215-288-5
Merck Index: 14,6260

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR