4-(Hydroxymethyl)phenylboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
L15194.03
L15194.06
L15194.14
L15194.03EA
52.1
EUR
L15194.03
L15194.06
L15194.14
4-(Hydroxymethyl)phenylboronsäure ≥98%
4-(Hydroxymethyl)phenylboronsäure
Purity: 98%
CAS No.: 59016-93-2
Molecular Formula: C7H9BO3
Formula Weight: 151.96
Melting Point: ca 280°
Beilstein Reference: 2831413
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...