3-Fluoro-DL-valine 94%

Lieferant: Thermo Scientific
Achtung

L15334.MC
L15334.MCEA 160 EUR
L15334.MC
3-Fluoro-DL-valine 94%
3-Fluoro-DL-valine

Purity: 94% CAS No.: 43163-94-6 Molecular Formula: C5H10FNO2 Formula Weight: 135.14 Melting Point: ca 250° dec. Beilstein Reference: 2350377 MDL No.: MFCD00077549

Formel: C₅H₁₀FNO₂
Molecular Weight: 135,14 g/mol
Schmelzpunkt: ∼ 250 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00077549
CAS-Nummer: 43163-94-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR