2-Brom-4'-(1-pyrrolidinyl)acetophenon ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-(Pyrrolidin-1-yl)phenacyl bromide
L15407.06
L15407.14
L15407.06EA
73.2
EUR
L15407.06
L15407.14
2-Brom-4'-(1-pyrrolidinyl)acetophenon ≥97%
2-Brom-4'-(1-pyrrolidinyl)acetophenon
Purity: 97%
CAS No.: 216144-18-2
Molecular Formula: C12H14BrNO
Formula Weight: 268.16
Melting Point: 120-122°
Beilstein Reference: 8549816
UN No.: 3261
MDL No.: MFCD01075690
Formel:
C₁₂H₁₄BrNO Schmelzpunkt: 120…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075690 CAS-Nummer: 216144-18-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...