Magnesium bromide diethyl etherate 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Magnesium bromide ethyl etherate

L15481.14
L15481.14EA 72.7 EUR
L15481.14
Magnesium bromide diethyl etherate 98%
Magnesium bromide diethyl etherate

Purity: 98% CAS No.: 29858-07-9 Molecular Formula: C4H10O.Br2Mg Formula Weight: 258.24 Flash Point: 35°(95°F) UN No.: UN1325 MDL No.: MFCD00064500

Formel: C₄H₁₀Br₂MgO
Molecular Weight: 258,236 g/mol
Flammpunkt: 35 °C (95 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064500
CAS-Nummer: 29858-07-9
ADR: 4.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR