3-Furaldehyde diethyl acetal 98%

Lieferant: Thermo Fisher Scientific
Gefahr

L15519.06
L15519.06EA 57.5 EUR
L15519.06
3-Furaldehyde diethyl acetal 98%
3-Furaldehyde diethyl acetal

Purity: 98% CAS No.: 216144-29-5 Molecular Formula: C9H14O3 Formula Weight: 170.21 Refractive Index: 1.4440 MDL No.: MFCD01318151

Formel: C₉H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01318151
CAS-Nummer: 216144-29-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR