Benzyl-tert-butyl malonate 95%
Lieferant: Thermo Scientific
Synonyme:
Benzyl tert-butyl malonate
L15533.06
L15533.06EA
71.3
EUR
L15533.06
Benzyl-tert-butyl malonate 95%
Benzyl-tert-butyl malonate
Purity: 95%
CAS No.: 72594-86-6
Molecular Formula: C14H18O4
Formula Weight: 250.30
Refractive Index: 1.4860
Beilstein Reference: 5434466
MDL No.: MFCD01075175
Formel:
C₁₄H₁₈O₄ Molecular Weight: 250,294 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075175 CAS-Nummer: 72594-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...