Cycloheptylacetic acid 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L15553.03
L15553.06
L15553.03EA
63.4
EUR
L15553.03
L15553.06
Cycloheptylacetic acid 99%
Cycloheptylacetic acid
Purity: 99%
CAS No.: 4401-20-1
Molecular Formula: C9H16O2
Formula Weight: 156.23
Refractive Index: 1.4725
Melting Point: 14-15°
Boiling Point: 110-112°/1mm
Beilstein Reference: 1935484
MDL No.: MFCD01075701
Formel:
C₉H₁₆O₂ Siedepunkt: 110…112 °C (1 mmHg) Schmelzpunkt: 14…15 °C Dichte: 1,023 Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075701 CAS-Nummer: 4401-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...