2,2'-(Methylimino)diethanol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-Methyldiethanolamin, N,N-Diethanolmethylamin, N-Methyl-2,2'-iminodiethanol, AMIETOL M 12, MDEA
L15712.36
L15712.22
L15712.36EA
53.6
EUR
L15712.36
L15712.22
2,2'-(Methylimino)diethanol ≥98%
2,2'-Methyliminodiethanol
Purity: 98+%
CAS No.: 105-59-9
Molecular Formula: C5H13NO2
Formula Weight: 119.17
Flash Point: 127°(260°F)
Refractive Index: 1.4690
Odour: Light
Melting Point: -21°
Boiling Point: 243°
Beilstein Reference: 1734441
MDL No.: MFCD00002848
Formel:
H₃CN(CH₂CH₂OH)₂ Siedepunkt: 243 °C Schmelzpunkt: –21 °C Dichte: 1,040 Flammpunkt: 127 °C (260 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002848 CAS-Nummer: 105-59-9 EINECS: 203-312-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...