6-Methoxy-1-indanon ≥99%

Lieferant: Thermo Fisher Scientific

L15819.06
L15819.06EA 90 EUR
L15819.06
6-Methoxy-1-indanon ≥99%
6-Methoxy-1-indanon

Purity: 99% CAS No.: 13623-25-1 Molecular Formula: C10H10O2 Formula Weight: 162.19 Melting Point: 104-109° Beilstein Reference: 1238602

Formel: C₁₀H₁₀O₂
Siedepunkt: 270 °C (1013 hPa)
Schmelzpunkt: 104…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00021232
CAS-Nummer: 13623-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR