Ethyl-4-amino-1-piperidincarboxylat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ethyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-piperidinecarboxylic Acid Ethyl Ester
L15866.06
L15866.06EA
32.2
EUR
L15866.06
Ethyl-4-amino-1-piperidincarboxylat 98%
Ethyl-4-amino-1-piperidinecarboxylate
Purity: 98%
CAS No.: 58859-46-4
Molecular Formula: C8H16N2O2
Formula Weight: 172.23
Flash Point: 78°(172°F)
Refractive Index: 1.4830
Boiling Point: 118-120°/6mm
Beilstein Reference: 388597
MDL No.: MFCD00006484
Formel:
C₈H₁₆N₂O₂ Molecular Weight: 172,23 g/mol Siedepunkt: 118…120 °C (6 mmHg) Dichte: 1,004 Flammpunkt: 78 °C (172 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006484 CAS-Nummer: 58859-46-4 EINECS: 261-472-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...