D-(-)-Pantolactone 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Pantolactone, (R)-(-)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, (R)-3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one, D-(-)-Pantoyl Lactone, D-(-)-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone
L15983.14
L15983.06
L15983.14EA
73.2
EUR
L15983.14
L15983.06
D-(-)-Pantolactone 99%
D-(-)-Pantolactone
Purity: 99%
CAS No.: 599-04-2
Molecular Formula: C6H10O3
Formula Weight: 130.14
Storage Temperature: 2°C to 8°C
Melting Point: 89-91°
Boiling Point: 120-122°/15mm
Beilstein Reference: 80957
Optical Rotation: -51° (c=3 in water)
MDL No.: MFCD00005392
Formel:
C₆H₁₀O₃ Siedepunkt: 120…122 °C (15 mmHg) Schmelzpunkt: 89…91 °C |
MDL:
MFCD00005392 CAS-Nummer: 599-04-2 EINECS: 209-963-3 Merck Index: 14,7013 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...