5-O-tert-Butyldiphenylsilyl-2,3-O-isopropylidene-α,β-D-ribofuranose
Lieferant: Thermo Scientific
L15989.06
L15989.06EA
330
EUR
L15989.06
5-O-tert-Butyldiphenylsilyl-2,3-O-isopropylidene-α,β-D-ribofuranose
5-O-tert-Butyldiphenylsilyl-2,3-O-isopropylidene-α,β-D-ribofuranose
CAS No.: 141607-35-4
Molecular Formula: C24H32O5Si
Formula Weight: 428.60
Storage Temperature: 2°C to 8°C
Optical Rotation: -28° (c=1.5 in chloroform)
MDL No.: MFCD01075719
Formel:
C₂₄H₃₂O₅Si |
MDL:
MFCD01075719 CAS-Nummer: 141607-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...