Methyl-pentafluorphenylcarbonat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L16107.06
L16107.06EA
77.5
EUR
L16107.06
Methyl-pentafluorphenylcarbonat 97%
Methyl-pentafluorphenylcarbonat
Purity: 97%
CAS No.: 36919-03-6
Molecular Formula: C8H3F5O3
Formula Weight: 242.10
Melting Point: 27-31°
Boiling Point: 84°/3mm
Beilstein Reference: 2289710
UN No.: UN2811
MDL No.: MFCD01075723
Formel:
C₈H₃F₅O₃ Siedepunkt: 84 °C (3 mmHg) Schmelzpunkt: 27…31 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075723 CAS-Nummer: 36919-03-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...