3-Bromothiophene-2-carboxamide 99%

Lieferant: Thermo Fisher Scientific
Achtung

L16138.03
L16138.03EA 145 EUR
L16138.03
3-Bromothiophene-2-carboxamide 99%
3-Bromothiophene-2-carboxamide

Purity: 99% CAS No.: 78031-18-2 Molecular Formula: C5H4BrNOS Formula Weight: 206.06 Melting Point: 106-108° Beilstein Reference: 4937660 MDL No.: MFCD00173835

Formel: C₅H₄BrNOS
Schmelzpunkt: 106…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173835
CAS-Nummer: 78031-18-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR