N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylendiamin 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,1',1'',1'''-(Ethane-1,2-diylbis(azanetriyl))tetrakis(propan-2-ol), Ethylenediamine-N,N,N',N'-tetra-2-propanol

L16280.AE L16280.AP
L16280.AEEA 30.4 EUR
L16280.AE L16280.AP
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylendiamin 99%
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylendiamin

Purity: 99% CAS No.: 102-60-3 Molecular Formula: C14H32N2O4 Formula Weight: 292.42 Flash Point: 110°(230°F) Refractive Index: 1.4800 Odour: Odourless Boiling Point: 240°/10mm Merck Reference: 14,3599 MDL No.: MFCD00004534

Formel: C₁₄H₃₂N₂O₄
Molecular Weight: 292.42 g/mol
Siedepunkt: 240 °C (10 mmHg)
Dichte: 1.030
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004534
CAS-Nummer: 102-60-3
EINECS: 203-041-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR