3-Methyl-4-isoxazolcarbonsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L16307.03
L16307.03EA
202
EUR
L16307.03
3-Methyl-4-isoxazolcarbonsäure ≥98%
3-Methyl-4-isoxazolecarboxylic acid
Purity: 98+%
CAS No.: 17153-20-7
Molecular Formula: C5H5NO3
Formula Weight: 127.10
Melting Point: 185-187°
Beilstein Reference: 114157
Formel:
C₅H₅NO₃ Schmelzpunkt: 185…187 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318161 CAS-Nummer: 17153-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...