5-Methylnicotinsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Methyl-3-pyridincarbonsäure, 5-Methylpyridin-3-carbonsäure

L16370.03 L16370.06
L16370.03EA 104 EUR
L16370.03 L16370.06
5-Methylnicotinsäure ≥97%
5-Methylnicotinsäure

Purity: 97% CAS No.: 3222-49-9 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Melting Point: 210-215° Beilstein Reference: 112288

Formel: C₇H₇NO₂
Schmelzpunkt: 210…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00829036
CAS-Nummer: 3222-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR