2,4-Dichlor-6-methylbenzonitril ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
L16372.06
L16372.06EA
136
EUR
L16372.06
2,4-Dichlor-6-methylbenzonitril ≥97%
2,4-Dichlor-6-methylbenzonitril
Purity: 97%
CAS No.: 175277-98-2
Molecular Formula: C8H5Cl2N
Formula Weight: 186.04
Melting Point: 79-81°
Beilstein Reference: 3249214
UN No.: 3439
MDL No.: MFCD00052865
Formel:
C₈H₅Cl₂N Molecular Weight: 186,04 g/mol Schmelzpunkt: 79…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052865 CAS-Nummer: 175277-98-2 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...