(3-(Methylamino)propyl)trimethoxysilan 95%

Lieferant: Thermo Fisher Scientific
Achtung

L16541.14 L16541.22
L16541.14EA 107 EUR
L16541.14 L16541.22
(3-(Methylamino)propyl)trimethoxysilan 95%
(3-(Methylamino)propyl)trimethoxysilan

Purity: 95% CAS No.: 3069-25-8 Molecular Formula: CH3NH(CH2)3Si(OCH3)3 Formula Weight: 193.32 Flash Point: 82°(179°F) Refractive Index: 1.4190 Boiling Point: 106°/30mm Beilstein Reference: 2072720 MDL No.: MFCD00048021

Formel: H₃CNHCH₂CH₂CH₂Si(OCH₃)₃
Siedepunkt: 106 °C (30 mmHg)
Dichte: 0,978
Flammpunkt: 82 °C (179 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00048021
CAS-Nummer: 3069-25-8
EINECS: 221-334-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR