2,2,3,3,4,4,5,5-Octafluor-1,6-hexandiol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L16560.06 L16560.14
L16560.06EA 55.5 EUR
L16560.06 L16560.14
2,2,3,3,4,4,5,5-Octafluor-1,6-hexandiol ≥97%
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol

Purity: 97% CAS No.: 355-74-8 Molecular Formula: HOCH2(CF2)4CH2OH Formula Weight: 262.10 Melting Point: 67-70° Boiling Point: 100°/3mm MDL No.: MFCD00069087

Formel: C₆H₆F₈O₂
Siedepunkt: 100 °C (3 mmHg)
Schmelzpunkt: 67…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00069087
CAS-Nummer: 355-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR