Dichlormethyloctylsilan ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L16565.09
L16565.18
L16565.09EA
19.6
EUR
L16565.09
L16565.18
Dichlormethyloctylsilan ≥98%
Dichlormethyloctylsilan
Purity: 98%
CAS No.: 14799-93-0
Molecular Formula: C9H20Cl2Si
Formula Weight: 227.25
Flash Point: 98°(208°F)
Refractive Index: 1.4435
Boiling Point: 224-226°
Beilstein Reference: 1740697
UN No.: 2987
MDL No.: MFCD00042606
Formel:
C₉H₂₀Cl₂Si Siedepunkt: 224…226 °C Dichte: 0,794 Flammpunkt: 98 °C (208 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042606 CAS-Nummer: 14799-93-0 EINECS: 238-863-2 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...