Pentamethyldisiloxan 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1,1,1,3,3-Pentamethyldisiloxane

L16577.06 L16577.14
L16577.06EA 51 EUR
L16577.06 L16577.14
Pentamethyldisiloxan 95%
Pentamethyldisiloxane

Purity: 95% CAS No.: 1438-82-0 Molecular Formula: (CH3)3SiOSiH(CH3)2 Formula Weight: 148.35 Flash Point: -19°(-2°F) Refractive Index: 1.3740 Boiling Point: 86-87° Beilstein Reference: 1734568 UN No.: 1993 MDL No.: MFCD00053658

Formel: C₅H₁₆OSi₂
Molecular Weight: 148.35 g/mol
Siedepunkt: 86…87 °C
Dichte: 0.758
Flammpunkt: –19 °C (–2 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00053658
CAS-Nummer: 1438-82-0
EINECS: 215-873-5
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR