3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl methacrylate, 3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane

L16682.14 L16682.22
L16682.14EA 87.7 EUR
L16682.14 L16682.22
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat ≥98%
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propylmethacrylat

Purity: 98% CAS No.: 17096-07-0 Molecular Formula: C16H38O5Si4 Formula Weight: 422.82 Flash Point: 201°(393°F) Refractive Index: 1.4175 Boiling Point: 112-115°/0.2mm MDL No.: MFCD00053871

Formel: C₁₆H₃₈O₅Si₄
Molecular Weight: 422.82 g/mol
Siedepunkt: 112…115 °C (0.2 mmHg)
Dichte: 0.930
Flammpunkt: 201 °C (393 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00053871
CAS-Nummer: 17096-07-0
EINECS: 241-165-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR