2-Brom-4,5-difluorphenol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Bromo-3,4-difluoro-6-hydroxybenzene

L16716.06
L16716.06EA 101 EUR
L16716.06
2-Brom-4,5-difluorphenol ≥97%
2-Brom-4,5-difluorphenol

Purity: 97% CAS No.: 166281-37-4 Molecular Formula: C6H3BrF2O Formula Weight: 208.99 Refractive Index: 1.5320 Beilstein Reference: 8355005

Formel: C₆H₃BrF₂O
Molecular Weight: 208.99 g/mol
Dichte: 1.829
Lagertemperatur: Raumtemperatur
MDL: MFCD00070751
CAS-Nummer: 166281-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR