N,N-Diethyl-2,3,3,3-tetrafluoropropionamide 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: N,N-Diethyl-2,3,3,3-tetrafluoropropanamide

L16739.14
L16739.14EA 74.2 EUR
L16739.14
N,N-Diethyl-2,3,3,3-tetrafluoropropionamide 95%
N,N-Diethyl-2,3,3,3-tetrafluoropropionamide

Purity: 95% CAS No.: 392-63-2 Molecular Formula: C7F4H11NO Formula Weight: 201.16 Refractive Index: 1.3920 Boiling Point: 58°/5mm Beilstein Reference: 1780406 MDL No.: MFCD00069160

Formel: C₇H₁₁F₄NO
Molecular Weight: 201,17 g/mol
Siedepunkt: 58 °C (5 mmHg)
Dichte: 1,228
Lagertemperatur: Raumtemperatur
MDL: MFCD00069160
CAS-Nummer: 392-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR